Please login to your online account to display your discounted pricing

4,5-Dichloro-2-methyl-3(2H)-pyridazinone, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC399710050

 View more versions of this product

Catalog No. AC399710050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 4, 5-Dichloro-2-methyl-3(2H)-pyridazinone
Quantity 5g
CAS 933-76-6
Assay Percent Range 97%
Molecular Formula C5H4Cl2N2O
Formula Weight 179.01
MDL Number MFCD00051686

Specifications

Chemical Name or Material 4, 5-Dichloro-2-methyl-3(2H)-pyridazinone
Assay Percent Range 97%
Molecular Formula C5H4Cl2N2O
Formula Weight 179.01
MDL Number MFCD00051686