Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Dichloro-2-(3,5-dichlorophenyl)-3(2H)-pyridazinone, 97%

$36.77 - $132.28

Chemical Identifiers

Chemical Name or Material 4,5-Dichloro-2-(3,5-dichlorophenyl)-3(2H)-pyridazinone
CAS 86483-43-4
Molecular Formula C10H4Cl4N2O
Formula Weight 309.97
MDL Number MFCD00067796
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2565103

  SDS
alfa aesar™
B2565103
1g Each for $36.77

AAB2565106

  SDS
alfa aesar™
B2565106
5g Each for $132.28
Description & Specifications

Specifications

Formula Weight 309.97
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00067796
Chemical Name or Material 4,5-Dichloro-2-(3,5-dichlorophenyl)-3(2H)-pyridazinone
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H4Cl4N2O
CAS 86483-43-4
Quantity 1g
Health Hazard 1 Warning
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.