Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Dichloro-1-methylimidazole, 97%

$104.11 - $394.77

Chemical Identifiers

Chemical Name or Material 4,5-Dichloro-1-methylimidazole
CAS 1192-53-6
Molecular Formula C4H4Cl2N2
Formula Weight 151
MDL Number MFCD00067911
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2015103

  SDS
alfa aesar™
B2015103
1g Each for $104.11

AAB2015106

  SDS
alfa aesar™
B2015106
5g Each for $394.77
Description & Specifications

Specifications

Formula Weight 151
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 1g
Chemical Name or Material 4,5-Dichloro-1-methylimidazole
Assay 97%
Molecular Formula C4H4Cl2N2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1192-53-6
Melting Point 53° to 55°C
MDL Number MFCD00067911

Applications
It is used as primary and secondary intermediate.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.