Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Dibromothiophene-2-carboxylic acid, 98%

$99.89 - $1,161.72

Chemical Identifiers

Chemical Name or Material 4,5-Dibromothiophene-2-carboxylic acid
CAS 6324-10-3
Molecular Formula C5H2Br2O2S
Formula Weight 285.94
MDL Number MFCD00039682
Beilstein 135885
Synonym 4,5-Dibromo-2-thenoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2092806

  SDS
alfa aesar™
B2092806
5g Each for $99.89

AAB2092814

  SDS
alfa aesar™
B2092814
25g Each for $363.14

AAB2092822

  SDS
alfa aesar™
B2092822
100g Each for $1,161.72
Description & Specifications

Specifications

Formula Weight 285.94
Chemical Name or Material 4,5-Dibromothiophene-2-carboxylic acid
Quantity 5g
Health Hazard 1 Warning
Melting Point 228° to 232°C
Assay 98%
Molecular Formula C5H2Br2O2S
Odor Odorless
MDL Number MFCD00039682
CAS 6324-10-3
Beilstein 135885
Synonym 4,5-Dibromo-2-thenoic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.