Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Dibromo-o-xylene, 97%

$62.65 - $247.82

Chemical Identifiers

Chemical Name or Material 4,5-Dibromo-o-xylene
CAS 24932-48-7
Molecular Formula C8H8Br2
Formula Weight 263.97
MDL Number MFCD00082743
Beilstein 2206183
Synonym 1,2-Dibromo-4,5-dimethylbenzene; 4,5-Dibromo-1,2-dimethylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2272703

  SDS
alfa aesar™
B2272703
1g Each for $62.65

AAB2272706

  SDS
alfa aesar™
B2272706
5g Each for $247.82
Description & Specifications

Specifications

Formula Weight 263.97
Chemical Name or Material 4,5-Dibromo-o-xylene
Assay 97%
Molecular Formula C8H8Br2
MDL Number MFCD00082743
CAS 24932-48-7
Beilstein 2206183
Synonym 1,2-Dibromo-4,5-dimethylbenzene; 4,5-Dibromo-1,2-dimethylbenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 85° to 87°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.