Learn More
4,5-Dibromo-3(2H)-pyridazinone, 98%, Thermo Scientific Chemicals
$230.10 - $801.08
Chemical Identifiers
CAS | 5788-58-9 |
---|---|
Molecular Formula | C4H2Br2N2O |
Molecular Weight (g/mol) | 253.881 |
MDL Number | MFCD00023641 |
InChI Key | AGLQURQNVJVJNB-UHFFFAOYSA-N |
Synonym | 4,5-dibromopyridazin-3 2h-one, 4,5-dibromo-3 2h-pyridazinone, 4,5-dibromopyridazin-3-one, 4,5-dibromopyridazine-3 2h-one, 4,5-dibromopyridazin-3-ol, 4,5-dibromo-2,3-dihydropyridazin-3-one, 3 2h-pyridazinone, 4,5-dibromo, 4,5-dibromo-2h-pyridazin-3-one, 4,5-dibromo-2-hydropyridazin-3-one |
PubChem CID | 236181 |
IUPAC Name | 4,5-dibromo-1H-pyridazin-6-one |
SMILES | C1=NNC(=O)C(=C1Br)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1599406
|
Thermo Scientific Chemicals
A1599406 |
5 g |
Each for $230.10
|
|
|||||
AAA1599414
|
Thermo Scientific Chemicals
A1599414 |
25 g |
Each for $801.08
|
|
|||||
Description
4,5-Dibromo-3(2H)-pyridazinone is used as an intermediate in the manufacture of pharmaceuticals. It is widely used in chemical research and in fine chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4,5-Dibromo-3(2H)-pyridazinone is used as an intermediate in the manufacture of pharmaceuticals. It is widely used in chemical research and in fine chemicals.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
5788-58-9 | |
253.881 | |
AGLQURQNVJVJNB-UHFFFAOYSA-N | |
236181 | |
C1=NNC(=O)C(=C1Br)Br |
C4H2Br2N2O | |
MFCD00023641 | |
4,5-dibromopyridazin-3 2h-one, 4,5-dibromo-3 2h-pyridazinone, 4,5-dibromopyridazin-3-one, 4,5-dibromopyridazine-3 2h-one, 4,5-dibromopyridazin-3-ol, 4,5-dibromo-2,3-dihydropyridazin-3-one, 3 2h-pyridazinone, 4,5-dibromo, 4,5-dibromo-2h-pyridazin-3-one, 4,5-dibromo-2-hydropyridazin-3-one | |
4,5-dibromo-1H-pyridazin-6-one |
Specifications
5788-58-9 | |
C4H2Br2N2O | |
5 g | |
4,5-dibromopyridazin-3 2h-one, 4,5-dibromo-3 2h-pyridazinone, 4,5-dibromopyridazin-3-one, 4,5-dibromopyridazine-3 2h-one, 4,5-dibromopyridazin-3-ol, 4,5-dibromo-2,3-dihydropyridazin-3-one, 3 2h-pyridazinone, 4,5-dibromo, 4,5-dibromo-2h-pyridazin-3-one, 4,5-dibromo-2-hydropyridazin-3-one | |
C1=NNC(=O)C(=C1Br)Br | |
253.881 | |
253.89 | |
4,5-Dibromo-3(2H)-pyridazinone |
∼220°C (decomposition) | |
MFCD00023641 | |
121469 | |
AGLQURQNVJVJNB-UHFFFAOYSA-N | |
4,5-dibromo-1H-pyridazin-6-one | |
236181 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only