missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Dibromo-2-fluorotoluene, 98%

Manufacturer:  Alfa Aesar™ H6332106

 View more versions of this product

Catalog No. AAH6332106



Specifications

Specifications

4,5-Dibromo-2-fluorotoluene
202982-77-2
MFCD00143234
GJHJZTCWLKJMIG-UHFFFAOYSA-N
1,2-dibromo-4-fluoro-5-methylbenzene
2773986
Solid
5g
C7H5Br2F
4,5-dibromo-2-fluorotoluene, 2-fluoro-4,5-dibromotoluene, pubchem10411, benzene,1,2-dibromo-4-fluoro-5-methyl, 1,2-bis bromanyl-4-fluoranyl-5-methyl-benzene
CC1=CC(=C(C=C1F)Br)Br
267.923
267.92
98%

Chemical Identifiers

202982-77-2
267.923
GJHJZTCWLKJMIG-UHFFFAOYSA-N
2773986
CC1=CC(=C(C=C1F)Br)Br
C7H5Br2F
MFCD00143234
4,5-dibromo-2-fluorotoluene, 2-fluoro-4,5-dibromotoluene, pubchem10411, benzene,1,2-dibromo-4-fluoro-5-methyl, 1,2-bis bromanyl-4-fluoranyl-5-methyl-benzene
1,2-dibromo-4-fluoro-5-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents