missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Diazafluoren-9-one, 98%

Manufacturer:  Alfa Aesar™ H5602803

 View more versions of this product

Catalog No. AAH5602803



Description

Description

Applications
4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].

Solubility
Soluble in dichloromethane, tetrahydrofuran, chloroform, benzene, toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.

Notes
Incompatible with strong oxidizing agents. Avoid heat and source of ignition.
Specifications

Specifications

4,5-Diazafluoren-9-one
1g
C11H6N2O
4,5-diaza-9h-fluoren-9-one
PFMTUGNLBQSHQC-UHFFFAOYSA-N
182.182
182.18
214°C to 217°C
50890-67-0
MFCD00046892
Soluble in dichloromethane,tetrahydrofuran,chloroform,benzene,toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.
C1=CC2=C(C3=C(C2=O)C=CC=N3)N=C1
342157
98%

Chemical Identifiers

50890-67-0
182.182
PFMTUGNLBQSHQC-UHFFFAOYSA-N
342157
C11H6N2O
MFCD00046892
4,5-diaza-9h-fluoren-9-one
C1=CC2=C(C3=C(C2=O)C=CC=N3)N=C1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents