Please login to your online account to display your discounted pricing

4-(5-Bromopyrimidin-2-yl)morpholine, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(5-Bromopyrimidin-2-yl)morpholine
CAS 84539-22-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C8H10BrN3O
Formula Weight 244.09
MDL Number MFCD00483251
Synonym 5-Bromo-2-(4-morpholino)pyrimidine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432700050

  SDS
thermo fisher scientific
AC432700050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC432700250

  SDS
thermo fisher scientific
AC432700250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0% (GC)
Formula Weight 244.09
Physical Form Crystalline Powder
Quantity 5g
Color White to off-white
Melting Point 86° to 90°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C8H10BrN3O
Assay Percent Range 97%
Chemical Name or Material 4-(5-Bromopyrimidin-2-yl)morpholine
MDL Number MFCD00483251
CAS 84539-22-0
Packaging Glass bottle
Synonym 5-Bromo-2-(4-morpholino)pyrimidine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.