missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,5-Bis(hydroxymethyl)imidazole, TCI America™

$299.45 - $1,032.80

Chemical Identifiers

CAS 33457-48-6
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.131
MDL Number MFCD00191661
InChI Key AGJXDKGTTMVHOU-UHFFFAOYSA-N
Synonym 1H-Imidazole-4,5-dimethanol
PubChem CID 573629
IUPAC Name [4-(hydroxymethyl)-1H-imidazol-5-yl]methanol
SMILES C1=NC(=C(N1)CO)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I04571G View Documents TCI America
I04571G
1g Each for $299.45
 
I04575G View Documents TCI America
I04575G
5g Each for $1,032.80
 
Specifications

Chemical Identifiers

33457-48-6
128.131
AGJXDKGTTMVHOU-UHFFFAOYSA-N
573629
C1=NC(=C(N1)CO)CO
C5H8N2O2
MFCD00191661
1H-Imidazole-4,5-dimethanol
[4-(hydroxymethyl)-1H-imidazol-5-yl]methanol

Specifications

White
1g
C5H8N2O2
1H-Imidazole-4,5-dimethanol
C1=NC(=C(N1)CO)CO
128.131
128.13
≥98.0% (T)
162°C
33457-48-6
MFCD00191661
AGJXDKGTTMVHOU-UHFFFAOYSA-N
[4-(hydroxymethyl)-1H-imidazol-5-yl]methanol
573629
Crystal-Powder at 20°C
4,5-Bis(hydroxymethyl)imidazole
Safety and Handling

Safety and Handling

TSCA : No