Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Bis(hydroxymethyl)-1-tritylimidazole, 97%

Chemical Identifiers

Chemical Name or Material 4,5-Bis(hydroxymethyl)-1-tritylimidazole
CAS 77134-74-8
Formula Weight 370.44
MDL Number MFCD03844709
Synonym 1-Trityl-4,5-imidazoledimethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH63533MD

 
alfa aesar™
H63533MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 370.44
Chemical Name or Material 4,5-Bis(hydroxymethyl)-1-tritylimidazole
Physical Form Solid
Assay 97%
MDL Number MFCD03844709
CAS 77134-74-8
Synonym 1-Trityl-4,5-imidazoledimethanol
Quantity 250mg
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C