Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene, 97%

$90.19 - $343.26

Chemical Identifiers

Chemical Name or Material 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene
CAS 161265-03-8
Molecular Formula C39H32OP2
Formula Weight 578.62
MDL Number MFCD00233866
Synonym Xantphos
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2624106

 
alfa aesar™
H2624106
5g Each for $343.26

AAH2624103

 
alfa aesar™
H2624103
1g Each for $90.19
Description & Specifications

Specifications

Formula Weight 578.62
Chemical Name or Material 4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene
Assay 97%
Molecular Formula C39H32OP2
MDL Number MFCD00233866
CAS 161265-03-8
Synonym Xantphos
Melting Point 224° to 228°C
Quantity 5g

Applications
4,5-Bis(diphenylphosphino)-9, 9-dimethylxanthene is used as a bidentate ligand, which finds application in hydroformylation of alkenes.

Solubility
Soluble in organic solvents.

Notes
Incompatible with strong oxidizing agents.