missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,5',8-Trimethylpsoralen, 99%, Thermo Scientific Chemicals

$161.00 - $625.58

Chemical Identifiers

CAS 3902-71-4
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00005010
InChI Key FMHHVULEAZTJMA-UHFFFAOYSA-N
Synonym trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
PubChem CID 5585
ChEBI CHEBI:28329
IUPAC Name 2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one
SMILES CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6322603
View Documents Promotion Details
Thermo Scientific Chemicals
J6322603
1 g
Each for $161.00
 
AAJ6322606
View Documents Promotion Details
Thermo Scientific Chemicals
J6322606
5 g
Each for $625.58
 
Description

Description

4,5',8-Trimethylpsoralen is used for treatment of psoriasis, and as a photochemical crosslinker of DNA used as a probe for nucleic acid structure, used to crosslink DNA onto mica surfaces, mutagenesis of N2 worms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5′,8-Trimethylpsoralen is used for treatment of psoriasis, and as a photochemical crosslinker of DNA used as a probe for nucleic acid structure, used to crosslink DNA onto mica surfaces, mutagenesis of N2 worms.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

3902-71-4
228.25
FMHHVULEAZTJMA-UHFFFAOYSA-N
5585
2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one
C14H12O3
MFCD00005010
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
CHEBI:28329
CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1

Specifications

3902-71-4
White
MFCD00005010
UN1759
14,9735
Soluble in chloroform
CC1=CC2=CC3=C(OC(=O)C=C3C)C(C)=C2O1
228.25
CHEBI:28329
99%
4,5',8-Trimethylpsoralen
229°C to 231°C
C14H12O3
1 g
221723
trioxsalen, trioxysalen, trisoralen, trimethylpsoralen, 4,5',8-trimethylpsoralen, elder 8011, trioxisaleno, trioxysalene, trioxysalenum, 2',4,8-trimethylpsoralen
FMHHVULEAZTJMA-UHFFFAOYSA-N
2,5,9-trimethyl-7H-furo[3,2-g]chromen-7-one
5585
228.25
Powder
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H351
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.

P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335-H351

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

EINECSNumber : 223-459-0

RTECSNumber : LV1576000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only