missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4',5,7-Trihydroxyflavanone, 97%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 139820100

Catalog No. AC139820100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4', 5, 7-Trihydroxyflavanone
480-41-1
100.0
96% min. (HPLC)
C15H12O5
10g
2r-naringenin, r-naringenin, naringenin, 2r-5,7-dihydroxy-2-4-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one, --naringenin, +-2r-5,7-dihydroxy-2-4-hydroxyphenyl chroman-4-one, 2,3-dihydro-5,7-dihydroxy-2-4-hydroxyphenyl-4-benzopyrone, +-naringenin, 2r-5,7,4'-trihydroxyflavone, 2r-4',5,7-trihydroxyflavanone
C1C(OC2=CC(=CC(=C2C1=O)O)O)C3=CC=C(C=C3)O
272.26
CHEBI:50201
97%
97%
96.0
Authentic
Glass bottle
MFCD00006844
15, 6506
FTVWIRXFELQLPI-CYBMUJFWSA-N
(2R)-5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
667495
272.26

Chemical Identifiers

480-41-1
272.26
FTVWIRXFELQLPI-CYBMUJFWSA-N
667495
(2R)-5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
C15H12O5
MFCD00006844
2r-naringenin, r-naringenin, naringenin, 2r-5,7-dihydroxy-2-4-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one, --naringenin, +-2r-5,7-dihydroxy-2-4-hydroxyphenyl chroman-4-one, 2,3-dihydro-5,7-dihydroxy-2-4-hydroxyphenyl-4-benzopyrone, +-naringenin, 2r-5,7,4'-trihydroxyflavone, 2r-4',5,7-trihydroxyflavanone
CHEBI:50201
C1C(OC2=CC(=CC(=C2C1=O)O)O)C3=CC=C(C=C3)O
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

EINECSNumber : 207-550-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only