missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4',5,7-Trihydroxyflavanone, 97%

Manufacturer:  Alfa Aesar™ L0983409

 View more versions of this product

Catalog No. AAL0983409



Description

Description

Applications
(±)-Naringenin is an antioxidant flavonoid. It has anti-inflammatory and antitumor properties. (±)-Naringenin induces apoptosis. It stimulates DNA repair following oxidative damage. (±)-Naringenin inhibits the activity of PI 3-kinase (PI(3)K).

Solubility
Soluble in ethanol (50 mg/ml), DMSO, methanol, and ammonium hydroxide (50 mg/ml). Insoluble in water (almost)

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

4',5,7-Trihydroxyflavanone
10g
C15H12O5
14,6424
FTVWIRXFELQLPI-UHFFFAOYSA-N
5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
932
272.26
248°C to 251°C
67604-48-2
MFCD00006844
naringenin, 5,7-dihydroxy-2-4-hydroxyphenyl chroman-4-one, naringenine, 4',5,7-trihydroxyflavanone, +/--naringenin, naringetol, salipurpol, --naringenin, narigenin, salipurol
C1C(OC2=CC(=CC(=C2C1=O)O)O)C3=CC=C(C=C3)O
272.256
CHEBI:50202
97%

Chemical Identifiers

67604-48-2
272.256
FTVWIRXFELQLPI-UHFFFAOYSA-N
932
5,7-dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
C15H12O5
MFCD00006844
naringenin, 5,7-dihydroxy-2-4-hydroxyphenyl chroman-4-one, naringenine, 4',5,7-trihydroxyflavanone, +/--naringenin, naringetol, salipurpol, --naringenin, narigenin, salipurol
CHEBI:50202
C1C(OC2=CC(=CC(=C2C1=O)O)O)C3=CC=C(C=C3)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DJ2981530

Documents