Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride, 96%

$149.18 - $498.74

Chemical Identifiers

Chemical Name or Material 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
CAS 28783-41-7
Molecular Formula C7H9NS·HCl
Formula Weight 175.68
MDL Number MFCD05861485
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2824303

  SDS
alfa aesar™
H2824303
1g Each for $149.18

AAH2824306

  SDS
alfa aesar™
H2824306
5g Each for $498.74
Description & Specifications

Specifications

Formula Weight 175.68
CAS 28783-41-7
Health Hazard 1 Warning
Melting Point 222° to 224°C
Assay 96%
Molecular Formula C7H9NS·HCl
MDL Number MFCD05861485
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Chemical Name or Material 4,5,6,7-Tetrahydrothieno[3,2-c]pyridine hydrochloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.