Please login to your online account to display your discounted pricing

4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde
CAS 307308-03-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H10N2O
Formula Weight 150.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC431090010

  SDS
thermo fisher scientific
AC431090010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC431090050

  SDS
thermo fisher scientific
AC431090050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Low Melting Solid or Liquid
Assay ≥96.0%
Formula Weight 150.18
Quantity 1g
Color White to yellow
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4,5,6,7-Tetrahydropyrazolo[1,5-a]pyridine-3-carbaldehyde
Assay Percent Range 97%
Molecular Formula C8H10N2O
CAS 307308-03-8
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.