Please login to your online account to display your discounted pricing

4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid
CAS 40133-07-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H10O2S
Formula Weight 182.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC438260050

 
thermo fisher scientific
AC438260050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.24
Assay ≥96.0%
Physical Form Crystalline Powder
Quantity 5g
Color White
Melting Point 195° to 205°C
CAS Max % 100.0
CAS Min % 96.0
Molecular Formula C9H10O2S
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4,5,6,7-Tetrahydrobenzo[b]thiophene-2-carboxylic acid
Assay Percent Range 97%
CAS 40133-07-1
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.