missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H64755MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4,5,6,7-Tetrahydro-5-azabenzimidazole hydrochloride | |
C6H10ClN3 | |
250 mg | |
4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl, 4,5,6,7-tetrahydro-1h-imidazo 4,5-c pyridine hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-5-azabenzimidazole hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloridyl, 1h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-1h-imidazo 3,4-c pyridine hydrochloride, 4,5,6,7-tetrahydro-imidazo-4,5-c-pyridine hydrochloride | |
C1CNCC2=C1N=CN2.Cl | |
159.617 | |
159.62 | |
Solid |
879668-17-4 | |
MFCD11846200 | |
15312806 | |
CHLFJSVTUQAGSC-UHFFFAOYSA-N | |
4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine;hydrochloride | |
21330723 | |
95% |
Chemical Identifiers
879668-17-4 | |
159.617 | |
CHLFJSVTUQAGSC-UHFFFAOYSA-N | |
21330723 | |
C1CNCC2=C1N=CN2.Cl |
C6H10ClN3 | |
MFCD11846200 | |
4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-3h-imidazo 4,5-c pyridine hcl, 4,5,6,7-tetrahydro-1h-imidazo 4,5-c pyridine hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-5-azabenzimidazole hydrochloride, 3h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloridyl, 1h,4h,5h,6h,7h-imidazo 4,5-c pyridine hydrochloride, 4,5,6,7-tetrahydro-1h-imidazo 3,4-c pyridine hydrochloride, 4,5,6,7-tetrahydro-imidazo-4,5-c-pyridine hydrochloride | |
4,5,6,7-tetrahydro-3H-imidazo[4,5-c]pyridine;hydrochloride |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only