Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5,6,7-Tetrahydro-1H-indazole, 98%

$28.47 - $75.57

Chemical Identifiers

Chemical Name or Material 4,5,6,7-Tetrahydro-1H-indazole
CAS 2305-79-5
Molecular Formula C7H10N2
Formula Weight 122.17
MDL Number MFCD00014565
Beilstein 2420
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0151503

  SDS
alfa aesar™
L0151503
1g Each for $28.47

AAL0151506

  SDS
alfa aesar™
L0151506
5g Each for $75.57
Description & Specifications

Specifications

Formula Weight 122.17
Chemical Name or Material 4,5,6,7-Tetrahydro-1H-indazole
Molecular Formula C7H10N2
MDL Number MFCD00014565
CAS 2305-79-5
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Beilstein 2420
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 140° to 142°C (2mmHg)
Melting Point 80° to 84°C
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C