Please login to your online account to display your discounted pricing

Alfa Aesar™ 4,5,6,7-Tetrachlorofluorescein

$10.65 - $10.65

Chemical Identifiers

Chemical Name or Material 4,5,6,7-Tetrachlorofluorescein
CAS 6262-21-1
Molecular Formula C20H8Cl4O5
Formula Weight 470.10g/mol
MDL Number MFCD00010030
Synonym 4,5,6,7-Tetrachloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]-xanthene]-3-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2563503

 
alfa aesar™
B25635-03
1g Each for $10.65
Description & Specifications

Specifications

Melting Point 304°C (dec.)
Quantity 1g
Chemical Name or Material 4,5,6,7-Tetrachlorofluorescein
CAS 6262-21-1
Molecular Formula C20H8Cl4O5
Formula Weight 470.10g/mol
MDL Number MFCD00010030
Synonym 4,5,6,7-Tetrachloro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-[9H]-xanthene]-3-one
TSCA TSCA