missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,5,6,7-Tetrabromobenzotriazole, TCI America™

Manufacturer:  TCI America T292010MG

 View more versions of this product

Catalog No. T292010MG



Specifications

Specifications

4,5,6,7-Tetrabromobenzotriazole
264°C
17374-26-4
MFCD06411399
OMZYUVOATZSGJY-UHFFFAOYSA-N
4,5,6,7-tetrabromo-2H-benzotriazole
1694
Crystal-Powder at 20°C
White-Yellow
10mg
C6HBr4N3
4,5,6,7-tetrabromobenzotriazole, tbb, tbbt, 4,5,6,7-tetrabromo-1h-benzotriazole, 4,5,6,7-tetrabromo-2-azabenzimidazole, ck2 inhibitor, casein kinase ii inhibitor i, 4,5,6,7-tetrabromo-1h-benzo d 1,2,3 triazole, tetrabromo-2-benzotriazole, 4,5,6,7-tetrabromo-1h-1,2,3-benzotriazole
C1(=C(C2=NNN=C2C(=C1Br)Br)Br)Br
434.711
434.71
≥97.0% (HPLC,T)

Chemical Identifiers

17374-26-4
434.711
OMZYUVOATZSGJY-UHFFFAOYSA-N
1694
C1(=C(C2=NNN=C2C(=C1Br)Br)Br)Br
C6HBr4N3
MFCD06411399
4,5,6,7-tetrabromobenzotriazole, tbb, tbbt, 4,5,6,7-tetrabromo-1h-benzotriazole, 4,5,6,7-tetrabromo-2-azabenzimidazole, ck2 inhibitor, casein kinase ii inhibitor i, 4,5,6,7-tetrabromo-1h-benzo d 1,2,3 triazole, tetrabromo-2-benzotriazole, 4,5,6,7-tetrabromo-1h-1,2,3-benzotriazole
4,5,6,7-tetrabromo-2H-benzotriazole
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents