Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[5-(1H-1,2,4-Triazol-1-ylmethyl)-1H-1,2,4-triazol-3-yl]pyridine

$123.00 - $123.00

Chemical Identifiers

Chemical Name or Material 4-[5-(1H-1,2,4-Triazol-1-ylmethyl)-1H-1,2,4-triazol-3-yl]pyridine
CAS 477852-54-3
Molecular Formula C10H9N7
Formula Weight 227.23
MDL Number MFCD08060950
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50644MD

 
alfa aesar™
H50644MD
250mg Each for $123.00
Description & Specifications

Specifications

Formula Weight 227.23
Chemical Name or Material 4-[5-(1H-1,2,4-Triazol-1-ylmethyl)-1H-1,2,4-triazol-3-yl]pyridine
Molecular Formula C10H9N7
MDL Number MFCD08060950
CAS 477852-54-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 250mg