Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[4-(Trifluoromethyl)phenyl]-3-thiosemicarbazide, 97%

Chemical Identifiers

Chemical Name or Material 4-[4-(Trifluoromethyl)phenyl]-3-thiosemicarbazide
CAS 206761-90-2
Molecular Formula C8H8F3N3S
Formula Weight 235.23
MDL Number MFCD00041301
UN Number UN2811
Beilstein 8061812
Synonym 4-(alpha,alpha,alpha-Trifluoro-p-tolyl)-3-thiosemicarbazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1159403

  SDS
alfa aesar™
L1159403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 235.23
CAS 206761-90-2
MDL Number MFCD00041301
Chemical Name or Material 4-[4-(Trifluoromethyl)phenyl]-3-thiosemicarbazide
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Beilstein 8061812
Melting Point 165° to 168°C
Assay 97%
Synonym 4-(alpha,alpha,alpha-Trifluoro-p-tolyl)-3-thiosemicarbazide
UN Number UN2811
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Molecular Formula C8H8F3N3S
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.