Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[4-(Trifluoromethoxy)phenoxy]piperidine, 97%

$119.45 - $415.61

Chemical Identifiers

Chemical Name or Material 4-[4-(Trifluoromethoxy)phenoxy]piperidine
CAS 287952-67-4
Molecular Formula C12H14F3NO2
Formula Weight 261.25
MDL Number MFCD06656166
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5081203

  SDS
alfa aesar™
H5081203
1g Each for $415.61

AAH50812MD

  SDS
alfa aesar™
H50812MD
250mg Each for $119.45
Description & Specifications

Specifications

Formula Weight 261.25
Chemical Name or Material 4-[4-(Trifluoromethoxy)phenoxy]piperidine
Assay 97%
Molecular Formula C12H14F3NO2
MDL Number MFCD06656166
CAS 287952-67-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 71° to 73°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.