missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-tert-Butylphenyl)benzoic Acid, TCI America™

$163.47 - $553.06

Chemical Identifiers

CAS 5748-42-5
Molecular Formula C17H18O2
Molecular Weight (g/mol) 254.329
MDL Number MFCD01631300
InChI Key HZIOPONJCVOCFE-UHFFFAOYSA-N
Synonym 4-tert-Butylbiphenyl-4′-carboxylic Acid
PubChem CID 541479
IUPAC Name 4-(4-tert-butylphenyl)benzoic acid
SMILES CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B19861G View Documents TCI America
B19861G
1g Each for $163.47
 
B19865G View Documents TCI America
B19865G
5g Each for $553.06
 
Specifications

Chemical Identifiers

5748-42-5
254.329
HZIOPONJCVOCFE-UHFFFAOYSA-N
541479
CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O
C17H18O2
MFCD01631300
4-tert-Butylbiphenyl-4′-carboxylic Acid
4-(4-tert-butylphenyl)benzoic acid

Specifications

White-Yellow
5748-42-5
MFCD01631300
HZIOPONJCVOCFE-UHFFFAOYSA-N
4-(4-tert-butylphenyl)benzoic acid
541479
Crystal-Powder at 20°C
4-(4-tert-Butylphenyl)benzoic Acid
1g
C17H18O2
4-tert-Butylbiphenyl-4′C-carboxylic Acid
CC(C)(C)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O
254.329
254.33
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No