Please login to your online account to display your discounted pricing

4-[4-(tert-Butyl)-1,3-thiazol-2-yl]benzaldehyde, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-[4-(tert-Butyl)-1, 3-thiazol-2-yl]benzaldehyde
CAS 175202-78-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H15NOS
Formula Weight 245.34
MDL Number MFCD00084914
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

KM04244DA

  SDS
fisher scientific company
KM04244DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.34
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-[4-(tert-Butyl)-1, 3-thiazol-2-yl]benzaldehyde
Assay Percent Range 97%
MDL Number MFCD00084914
Packaging Amber Glass Bottle
CAS 175202-78-5
Molecular Formula C14H15NOS