Please login to your online account to display your discounted pricing

4-[4-(tert-Butyl)-1,3-thiazol-2-yl]benzaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  KM04244DA

 View more versions of this product

Catalog No. KM04244DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 245.34
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 4-[4-(tert-Butyl)-1, 3-thiazol-2-yl]benzaldehyde
Assay Percent Range 97%
MDL Number MFCD00084914
Packaging Amber Glass Bottle
CAS 175202-78-5
Molecular Formula C14H15NOS

Specifications

Chemical Name or Material 4-[4-(tert-Butyl)-1, 3-thiazol-2-yl]benzaldehyde
CAS 175202-78-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H15NOS
Formula Weight 245.34
MDL Number MFCD00084914