Please login to your online account to display your discounted pricing

4-[4-(tert-Butoxycarbonyl)piperazino]benzoic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC30001CB

 View more versions of this product

Catalog No. CC30001CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 306.36
Quantity 250mg
Assay 97%
Chemical Name or Material 4-[4-(tert-Butoxycarbonyl)piperazino]benzoic acid
CAS 162046-66-4
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C16H22N2O4

Specifications

Chemical Name or Material 4-[4-(tert-Butoxycarbonyl)piperazino]benzoic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C16H22N2O4
Formula Weight 306.36