Learn More
4-(4-Pyridylmethyl)aniline, 97%, Thermo Scientific™
Manufacturer: Fisher Scientific Company BTB01214CB
Specifications
4-(4-Pyridylmethyl)aniline | |
96.5 | |
97% | |
MFCD00053060 | |
4-4-pyridinylmethyl aniline, 4-4-pyridylmethyl aniline, 4-4-pyridylmethyl phenylamine, 4-p-aminobenzylpyridine, 4-pyridin-4-yl methyl aniline, 4-pyridin-4-ylmethyl aniline, 4-pyridin-4-ylmethyl phenyl amine, 4-pyridin-4-ylmethyl-phenylamine, benzenamine, 4-4-pyridinylmethyl, enamine_005904 | |
WZXYYQHVDJMIFF-UHFFFAOYSA-N | |
4-(pyridin-4-ylmethyl)aniline | |
307158 | |
97% |
27692-74-6 | |
100.0 | |
C12H12N2 | |
Amber Glass Bottle | |
250mg | |
C1=CC(=CC=C1CC2=CC=NC=C2)N | |
184.242 | |
184.24 | |
Technical |
Chemical Identifiers
27692-74-6 | |
184.242 | |
WZXYYQHVDJMIFF-UHFFFAOYSA-N | |
307158 | |
C1=CC(=CC=C1CC2=CC=NC=C2)N |
C12H12N2 | |
MFCD00053060 | |
4-4-pyridinylmethyl aniline, 4-4-pyridylmethyl aniline, 4-4-pyridylmethyl phenylamine, 4-p-aminobenzylpyridine, 4-pyridin-4-yl methyl aniline, 4-pyridin-4-ylmethyl aniline, 4-pyridin-4-ylmethyl phenyl amine, 4-pyridin-4-ylmethyl-phenylamine, benzenamine, 4-4-pyridinylmethyl, enamine_005904 | |
4-(pyridin-4-ylmethyl)aniline |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning