missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(4-Pyridyl)morpholine, TCI America™

Manufacturer:  TCI America P164625G

 View more versions of this product

Catalog No. P164625G



Specifications

Specifications

4-(4-Pyridyl)morpholine
107°C
2767-91-1
4-morpholinopyridine, 4-pyridin-4-yl morpholine, 4-4-pyridyl morpholine, morpholine, 4-4-pyridinyl, acmc-1cmri, 4-4-pyridinyl morpholine, 4-4-morpholinyl pyridine, 4-morpholinopyridine, 4-4-morpholinyl pyridine
C1COCCN1C2=CC=NC=C2
164.208
164.21
≥98.0% (GC,T)
Undesignated
25g
C9H12N2O
QJWQYVJVCXMTJP-UHFFFAOYSA-N
4-pyridin-4-ylmorpholine
4145413
Crystal-Powder at 20°C

Chemical Identifiers

2767-91-1
164.208
4-morpholinopyridine, 4-pyridin-4-yl morpholine, 4-4-pyridyl morpholine, morpholine, 4-4-pyridinyl, acmc-1cmri, 4-4-pyridinyl morpholine, 4-4-morpholinyl pyridine, 4-morpholinopyridine, 4-4-morpholinyl pyridine
4-pyridin-4-ylmorpholine
C9H12N2O
QJWQYVJVCXMTJP-UHFFFAOYSA-N
4145413
C1COCCN1C2=CC=NC=C2
Safety and Handling

Safety and Handling

TSCA : No

Documents