missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Pyridyl)benzaldehyde, 97%

Manufacturer:  Alfa Aesar™ H5546203

 View more versions of this product

Catalog No. AAH5546203



Specifications

Specifications

4-(4-Pyridyl)benzaldehyde
1g
C12H9NO
4-4-formylphenyl pyridine
C1=CC(=CC=C1C=O)C2=CC=NC=C2
183.21
183.21
85°C to 89°C
99163-12-9
MFCD02684110
MBJXDIYHLGBQOT-UHFFFAOYSA-N
4-pyridin-4-ylbenzaldehyde
639673
97%

Chemical Identifiers

99163-12-9
183.21
MBJXDIYHLGBQOT-UHFFFAOYSA-N
639673
C1=CC(=CC=C1C=O)C2=CC=NC=C2
C12H9NO
MFCD02684110
4-4-formylphenyl pyridine
4-pyridin-4-ylbenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents