missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Pyridyl)-2-thiazoleacetonitrile, 97%

$77.88 - $248.28

Chemical Identifiers

CAS 1016888-78-0
Molecular Formula C10H7N3S
Molecular Weight (g/mol) 201.247
MDL Number MFCD09950233
InChI Key LIIDMCZQVRSSQE-UHFFFAOYSA-N
Synonym 4-4-pyridyl thiazole-2-yl acetonitrile, 2-4-pyridin-4-yl thiazol-2-yl acetonitrile, 2-4-pyridin-4-yl-1,3-thiazol-2-yl acetonitrile
PubChem CID 24710447
IUPAC Name 2-(4-pyridin-4-yl-1,3-thiazol-2-yl)acetonitrile
SMILES C1=CN=CC=C1C2=CSC(=N2)CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65261MD View Documents Alfa Aesar™
H65261MD
250mg Each for $77.88
 
AAH6526103 View Documents Alfa Aesar™
H6526103
1g Each for $248.28
 
Specifications

Chemical Identifiers

1016888-78-0
201.247
LIIDMCZQVRSSQE-UHFFFAOYSA-N
24710447
C1=CN=CC=C1C2=CSC(=N2)CC#N
C10H7N3S
MFCD09950233
4-4-pyridyl thiazole-2-yl acetonitrile, 2-4-pyridin-4-yl thiazol-2-yl acetonitrile, 2-4-pyridin-4-yl-1,3-thiazol-2-yl acetonitrile
2-(4-pyridin-4-yl-1,3-thiazol-2-yl)acetonitrile

Specifications

Yellow to Brown
1016888-78-0
MFCD09950233
4-4-pyridyl thiazole-2-yl acetonitrile, 2-4-pyridin-4-yl thiazol-2-yl acetonitrile, 2-4-pyridin-4-yl-1,3-thiazol-2-yl acetonitrile
C1=CN=CC=C1C2=CSC(=N2)CC#N
201.247
201.25
97%
250mg
C10H7N3S
UN3439
LIIDMCZQVRSSQE-UHFFFAOYSA-N
2-(4-pyridin-4-yl-1,3-thiazol-2-yl)acetonitrile
24710447
Solid
4-(4-Pyridyl)-2-thiazoleacetonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.