missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(4-Phenylbutoxy)benzoic Acid 98.0+%, TCI America™
Supplier: TCI America P199725G
Specifications
4-(4-Phenylbutoxy)benzoic Acid | |
131°C | |
C17H18O3 | |
25 g | |
OC(=O)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1 | |
270.33 | |
270.33 | |
Crystalline Powder |
30131-16-9 | |
White | |
MFCD07787608 | |
XWCWFMQMZZPALR-UHFFFAOYSA-N | |
4-(4-phenylbutoxy)benzoic acid | |
11043825 | |
≥98.0% (GC,T) |
Chemical Identifiers
30131-16-9 | |
270.33 | |
XWCWFMQMZZPALR-UHFFFAOYSA-N | |
4-(4-phenylbutoxy)benzoic acid |
C17H18O3 | |
MFCD07787608 | |
11043825 | |
OC(=O)C1=CC=C(OCCCCC2=CC=CC=C2)C=C1 |
Safety and Handling
TSCA : No