Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Phenyl-1H-pyrazol-1-yl)piperidine, 95%

$96.97 - $346.34

Chemical Identifiers

Chemical Name or Material 4-(4-Phenyl-1H-pyrazol-1-yl)piperidine
CAS 902836-40-2
Molecular Formula C14H17N3
Formula Weight 227.31
MDL Number MFCD08060998
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5080503

  SDS
alfa aesar™
H5080503
1g Each for $346.34

AAH50805MD

  SDS
alfa aesar™
H50805MD
250mg Each for $96.97
Description & Specifications

Specifications

Formula Weight 227.31
Health Hazard 1 Warning
Chemical Name or Material 4-(4-Phenyl-1H-pyrazol-1-yl)piperidine
Assay 95%
Molecular Formula C14H17N3
MDL Number MFCD08060998
CAS 902836-40-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
4-(4-Phenyl-1H-pyrazol-1-yl)piperidine is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.