missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4-Pentamethylene-2-pyrrolidone, TCI America™

Manufacturer:  TCI America P16955G

 View more versions of this product

Catalog No. P16955G



Specifications

Specifications

4,4-Pentamethylene-2-pyrrolidone
White-Yellow
5g
≥98.0% (GC)
MFCD00177938
JAWPQJDOQPSNIQ-UHFFFAOYSA-N
2-azaspiro[4.5]decan-3-one
47457
Crystal-Powder at 20°C
181°C
91°C
64744-50-9
C9H15NO
3-Azaspiro[4.5]decan-2-one
C1CCC2(CC1)CC(=O)NC2
153.225
153.23

Chemical Identifiers

64744-50-9
153.225
JAWPQJDOQPSNIQ-UHFFFAOYSA-N
47457
C1CCC2(CC1)CC(=O)NC2
C9H15NO
MFCD00177938
3-Azaspiro[4.5]decan-2-one
2-azaspiro[4.5]decan-3-one
Safety and Handling

Safety and Handling

RTECSNumber : CM0120000

TSCA : No

Documents