missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4,4-Pentamethylene-2-pyrrolidone 98.0+%, TCI America™
Supplier: TCI America P169525G
Specifications
4,4-Pentamethylene-2-pyrrolidone | |
91°C | |
181°C | |
MFCD00177938 | |
3-Azaspiro[4.5]decan-2-one | |
O=C1CC2(CN1)CCCCC2 | |
153.23 | |
153.23 | |
Crystalline Powder |
64744-50-9 | |
White-Yellow | |
C9H15NO | |
25 g | |
JAWPQJDOQPSNIQ-UHFFFAOYSA-N | |
2-azaspiro[4.5]decan-3-one | |
47457 | |
≥98.0% (GC) |
Chemical Identifiers
64744-50-9 | |
153.23 | |
JAWPQJDOQPSNIQ-UHFFFAOYSA-N | |
47457 | |
O=C1CC2(CN1)CCCCC2 |
C9H15NO | |
MFCD00177938 | |
3-Azaspiro[4.5]decan-2-one | |
2-azaspiro[4.5]decan-3-one |
Safety and Handling
RTECSNumber : CM0120000
TSCA : No