Please login to your online account to display your discounted pricing

4,4'-Oxydiphenol, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4, 4';-Oxydiphenol
CAS 1965-09-9
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H10O3
Linear Formula HOC6H4OC6H4OH
Formula Weight 202.21
MDL Number MFCD00016463
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC416680250

  SDS
thermo fisher scientific
416680250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 202.21
Physical Form Amorphous Powder
Quantity 25g
Color Off-white
Melting Point 163° to 168°C
CAS Max % 100.0
Chemical Name or Material 4, 4';-Oxydiphenol
Assay Percent Range 99%
Molecular Formula C12H10O3
CAS 1965-09-9
MDL Number MFCD00016463
CAS Min % 98.5
Packaging Glass bottle
Linear Formula HOC6H4OC6H4OH
Infrared Spectrum Authentic