missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4,4'-Oxydiphenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 416680250

 View more versions of this product

Catalog No. AC416680250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4, 4';-Oxydiphenol
98.5
Authentic
≥98.5%
C12H10O3
202.21
Glass bottle
163° to 168°C
1965-09-9
100.0
Amorphous Powder
99%
HOC6H4OC6H4OH
MFCD00016463
Off-white
25g

Chemical Identifiers

1965-09-9
100.0
C12H10O3
202.21
4, 4';-Oxydiphenol
98.5
99%
HOC6H4OC6H4OH
MFCD00016463
≥98.5%