Please login to your online account to display your discounted pricing

4-(4-Nitrophenylazo)-1-naphthol, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  195750050

 View more versions of this product

Catalog No. AC195750050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 293.27
Quantity 5g
Color Red-brown
EINECS Number 226-129-4
TSCA TSCA
RTECS Number QL4510100
Beilstein 16, 155
CAS Max % 100.0
Chemical Name or Material 4-(4-Nitrophenylazo)-1-naphthol
Packaging Glass bottle
UV_Vis 98.5% min.
Name Note 99%
Assay Percent Range 99%
Molecular Formula C16H11N3O3
MDL Number MFCD00003975
CAS 5290-62-0
CAS Min % 98.5
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-(4-Nitrophenylazo)-1-naphthol
Name Note 99%
CAS 5290-62-0
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C16H11N3O3
Formula Weight 293.27
MDL Number MFCD00003975