Please login to your online account to display your discounted pricing

4-(4-Nitrophenylazo)-1-naphthol, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-(4-Nitrophenylazo)-1-naphthol
Name Note 99%
CAS 5290-62-0
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C16H11N3O3
Formula Weight 293.27
MDL Number MFCD00003975
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC195750050

  SDS
thermo fisher scientific
195750050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 293.27
Quantity 5g
Color Red-brown
TSCA TSCA
RTECS Number QL4510100
EINECS Number 226-129-4
Beilstein 16, 155
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 98.5
UV_Vis 98.5% min.
Packaging Glass bottle
Chemical Name or Material 4-(4-Nitrophenylazo)-1-naphthol
MDL Number MFCD00003975
Assay Percent Range 99%
Molecular Formula C16H11N3O3
Name Note 99%
CAS 5290-62-0

Indicator