Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Nitrophenylazo)-1-naphthol

$44.01 - $598.78

Chemical Identifiers

Chemical Name or Material 4- (4-Nitrophenylazo)-1-naphthol
CAS 5290-62-0
Molecular Formula C16H11N3O3
Formula Weight 293.28
MDL Number MFCD00003975
Beilstein 961660
Synonym Magneson II
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1127709

 
alfa aesar™
A1127709
10g Each for $44.01

AAA1127718

 
alfa aesar™
A1127718
50g Each for $141.76

AAA1127730

 
alfa aesar™
A1127730
250g Each for $598.78
Description & Specifications

Specifications

Formula Weight 293.28
Chemical Name or Material 4- (4-Nitrophenylazo)-1-naphthol
TSCA TSCA
Molecular Formula C16H11N3O3
MDL Number MFCD00003975
CAS 5290-62-0
Beilstein 961660
Synonym Magneson II
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number QL4510100
Melting Point ≈270°C (dec.)
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.