missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Nitrophenylazo)-1-naphthol

Manufacturer:  Alfa Aesar™ A1127730

 View more versions of this product

Catalog No. AAA1127730



Specifications

Specifications

4- (4-Nitrophenylazo)-1-naphthol
250g
C16H11N3O3
961660
UYDXOFSMBKSLPM-OBGWFSINSA-N
(4E)-4-[(4-nitrophenyl)hydrazinylidene]naphthalen-1-one
5918035
~270°C (decomposition)
5290-62-0
MFCD00003975
Magneson II
C1=CC=C2C(=O)C=CC(=NNC3=CC=C(C=C3)[N+](=O)[O-])C2=C1
293.282
293.28

Chemical Identifiers

5290-62-0
293.282
UYDXOFSMBKSLPM-OBGWFSINSA-N
5918035
C1=CC=C2C(=O)C=CC(=NNC3=CC=C(C=C3)[N+](=O)[O-])C2=C1
C16H11N3O3
MFCD00003975
Magneson II
(4E)-4-[(4-nitrophenyl)hydrazinylidene]naphthalen-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QL4510100

TSCA : TSCA

SDS Documents