Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Nitrophenyl)morpholine, 98%

$58.81 - $494.11

Chemical Identifiers

Chemical Name or Material 4-(4-Nitrophenyl)morpholine
CAS 10389-51-2
Molecular Formula C10H12N2O3
Formula Weight 208.22
MDL Number MFCD00023317
Synonym 1-(4-Morpholinyl)-4-nitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2493306

 
alfa aesar™
B2493306
5g Each for $58.81

AAB2493314

 
alfa aesar™
B2493314
25g Each for $164.70

AAB2493322

 
alfa aesar™
B2493322
100g Each for $494.11
Description & Specifications

Specifications

Formula Weight 208.22
Chemical Name or Material 4-(4-Nitrophenyl)morpholine
Assay 98%
Molecular Formula C10H12N2O3
MDL Number MFCD00023317
CAS 10389-51-2
Synonym 1-(4-Morpholinyl)-4-nitrobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number QE7405000
Melting Point 148° to 152°C
Quantity 5g
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.