Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Nitrophenyl)-1,2,3-thiadiazole, 97%

$65.99 - $65.99

Chemical Identifiers

Chemical Name or Material 4-(4-Nitrophenyl)-1,2,3-thiadiazole
CAS 82894-98-2
Molecular Formula C8H5N3O2S
Formula Weight 207.21
MDL Number MFCD00052103
Beilstein 982670
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1508503

  SDS
alfa aesar™
A1508503
1g Each for $65.99
Description & Specifications

Specifications

Formula Weight 207.21
Chemical Name or Material 4-(4-Nitrophenyl)-1,2,3-thiadiazole
Assay 97%
Molecular Formula C8H5N3O2S
MDL Number MFCD00052103
CAS 82894-98-2
Beilstein 982670
Melting Point 208° to 210°C
Quantity 1g