Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(4-Nitrobenzyloxy)acetophenone, 97%

$29.69 - $357.11

Chemical Identifiers

Chemical Name or Material 4'-(4-Nitrobenzyloxy)acetophenone
CAS 205748-03-4
Molecular Formula C15H13NO4
Formula Weight 271.28
MDL Number MFCD00075690
Beilstein 8321694
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1673803

 
alfa aesar™
A1673803
1g Each for $29.69

AAA1673806

 
alfa aesar™
A1673806
5g Each for $93.73

AAA1673814

 
alfa aesar™
A1673814
25g Each for $357.11
Description & Specifications

Specifications

Formula Weight 271.28
Melting Point 129° to 131°C
Assay 97%
Molecular Formula C15H13NO4
Quantity 1g
Beilstein 8321694
CAS 205748-03-4
Chemical Name or Material 4'-(4-Nitrobenzyloxy)acetophenone
MDL Number MFCD00075690