Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Nitrobenzyl)pyridine, 98+%

$135.46 - $135.46

Chemical Identifiers

Chemical Name or Material 4-(4-Nitrobenzyl)pyridine
CAS 1083-48-3
Molecular Formula C12H10N2O2
Formula Weight 214.22
MDL Number MFCD00006445
Beilstein 170877
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1356414

 
alfa aesar™
A1356414
25g Each for $135.46
Description & Specifications

Specifications

Formula Weight 214.22
Chemical Name or Material 4-(4-Nitrobenzyl)pyridine
TSCA TSCA
Assay ≥98%
Molecular Formula C12H10N2O2
MDL Number MFCD00006445
CAS 1083-48-3
Beilstein 170877
RTECS Number UT6360000
Melting Point 69° to 72°C
Quantity 25g

Applications
4-(4-Nitrobenzyl)pyridine is used in spectroscopic method for the determination of phosgene. It is also used in tests for alkylating agents.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong acids.