Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester, 97%

$177.01 - $603.39

Chemical Identifiers

Chemical Name or Material 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester
CAS 364794-79-6
Molecular Formula C17H26BNO3
Formula Weight 303.2
MDL Number MFCD04974052
Synonym 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2803403

 
alfa aesar™
H2803403
1g Each for $177.01

AAH2803406

 
alfa aesar™
H2803406
5g Each for $603.39
Description & Specifications

Specifications

Formula Weight 303.2
Chemical Name or Material 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C17H26BNO3
MDL Number MFCD04974052
CAS 364794-79-6
Synonym 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 86° to 89°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.