Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester, 97%

$101.26 - $323.96

Chemical Identifiers

Chemical Name or Material 4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester
CAS 656239-38-2
Molecular Formula C17H24BNO4
Formula Weight 317.19
MDL Number MFCD05863913
Synonym 4-(Morpholine-4-carbonyl)phenylboronic acid pinacol ester; 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2848403

  SDS
alfa aesar™
H2848403
1g Each for $101.26

AAH2848406

  SDS
alfa aesar™
H2848406
5g Each for $323.96
Description & Specifications

Specifications

Formula Weight 317.19
Chemical Name or Material 4-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester
Assay 97%
Molecular Formula C17H24BNO4
MDL Number MFCD05863913
CAS 656239-38-2
Synonym 4-(Morpholine-4-carbonyl)phenylboronic acid pinacol ester; 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
Melting Point 187° to 193°C
Quantity 1g