missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(4-Morpholinylcarbonyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H53126MD

 View more versions of this product

Catalog No. AAH53126MD



Specifications

Specifications

4-(4-Morpholinylcarbonyl)benzeneboronic acid
250mg
C11H14BNO4
4-morpholine-4-carbonyl phenylboronic acid, 4-morpholine-4-carbonyl phenyl boronic acid, 4-morpholinocarbonyl phenyl boronic acid, 4-morpholine-4-carbonyl phenyl boronic acid, 4-4-boronobenzoyl morpholine, 4-morpholine-4-carbonyl benzeneboronic acid, boronic acid, 4-4-morpholinylcarbonyl phenyl, 4-boronophenyl morpholin-4-yl methanone, 4-morpholin-4-ylcarbonyl benzeneboronic acid, boronic acid,b-4-4-morpholinylcarbonyl phenyl
B(C1=CC=C(C=C1)C(=O)N2CCOCC2)(O)O
235.046
235.05
126°C to 132°C
389621-84-5
MFCD03411952
KMNLIQJXZPBCDU-UHFFFAOYSA-N
[4-(morpholine-4-carbonyl)phenyl]boronic acid
2773546
98%

Chemical Identifiers

389621-84-5
235.046
KMNLIQJXZPBCDU-UHFFFAOYSA-N
2773546
B(C1=CC=C(C=C1)C(=O)N2CCOCC2)(O)O
C11H14BNO4
MFCD03411952
4-morpholine-4-carbonyl phenylboronic acid, 4-morpholine-4-carbonyl phenyl boronic acid, 4-morpholinocarbonyl phenyl boronic acid, 4-morpholine-4-carbonyl phenyl boronic acid, 4-4-boronobenzoyl morpholine, 4-morpholine-4-carbonyl benzeneboronic acid, boronic acid, 4-4-morpholinylcarbonyl phenyl, 4-boronophenyl morpholin-4-yl methanone, 4-morpholin-4-ylcarbonyl benzeneboronic acid, boronic acid,b-4-4-morpholinylcarbonyl phenyl
[4-(morpholine-4-carbonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents